Sulpho-LC-SDA (Sulpho-NHS-LC-Diazirine) (Sulphosuccinimidyl 6-(4,4′-azipentanamido)hexanoate), Pierce™
PIER26174
:
- Pk:50 mg
- Seq:2370
- Product Line:Pierce™
- Labeling Method:Chemical Labeling
- Crosslinker Type:Heterobifunctional
- Reactive Moiety:Diazirine, Sulfo-NHS Ester
- Spacer:Long
- Form:Solid
- Quantity:50 mg
- Type:Crosslinker
- Solubility:Water
- Spacer Arm:12,5 Å
:
:440.40 g/mol
:Refrigerator
:C₁₅H₂₁N₄NaO₈S
:MFCD28899432