SDA (NHS-Diazirine) (succinimidyl 4,4′-azipentanoate), Pierce™
Catalog # PIER26167
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:50 mg
- Seq:2370
Specifications
Specification Test Results
- Product Line:Pierce™
- Labeling Method:Chemical Labeling
- Crosslinker Type:Heterobifunctional
- Reactive Moiety:NHS Ester, Diazirine
- Spacer:Short
- Form:Solid
- Quantity:50 mg
- Type:Crosslinker
- Solubility:DMF, DMS
- Spacer Arm:3.9 Å
Specification Test Results
About this item
:
:225.20 g/mol
Storage Temperature:Refrigerator
Formula:C₉H₁₁N₃O₄
MDL Number:MFCD28899672