Order Entry
Northern Ireland
ContactUsLinkComponent
 
Fluphenazine Hydrochloride, Reference Standard
  1282004.
 :  USP
 :  
Fluphenazine Hydrochloride, Reference Standard
  1282004.
 :  USP
 :  

 

  • Pack type:
    Vial
  • Pk:
    125 mg
  • Standards:
    USP reference standard
  • CAS No.:
    146-56-5
  • Description:
    Fluphenazine Hydrochloride
  • UN No.:
    2811
  • Description:
    Fluphenazine Hydrochloride

 

 

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

 : US Pharmacopea Reference Standard