Order Entry
Northern Ireland
ContactUsLinkComponent
 
Chloropropyl Vanillin, Reference Standard
Catalog # 1117110.
Supplier:  USP
CAS Number:  
Chloropropyl Vanillin, Reference Standard
Catalog # 1117110.
Supplier:  USP
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Vial
  • Pk:
    25 mg
  • Standards:
    USP reference standard
  • CAS No.:
    151719-92-5
  • Description:
    Chloropropyl Vanillin
  • Synonym(s):
    4-(3-Chloropropoxy)-3-methoxybenzaldehyde
  • Description:
    Chloropropyl Vanillin

Specifications

About this item

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard