Order Entry
Northern Ireland
ContactUsLinkComponent
Acesulfame Potassium, Reference Standard
Acesulfame Potassium, Reference Standard
Catalog # 1002505.
Supplier:  USP
CAS Number:  
Acesulfame Potassium, Reference Standard
Catalog # 1002505.
Supplier:  USP
CAS Number:  

Specifications

  • Pk:
    200 mg
  • Standards:
    USP reference standard
  • CAS No.:
    55589-62-3
  • Description:
    Acesulfame Potassium
  • Storage conditions:
    Ambient
  • Formula:
    C₄H₄KNO₄S
  • Description:
    Acesulfame Potassium

Specifications

About this item

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard