Order Entry
Northern Ireland
ContactUsLinkComponent
Sulpho-SIAB (Sulphosuccinimidyl (4-iodoacetyl)aminobenzoate)
Sulpho-SIAB (Sulphosuccinimidyl (4-iodoacetyl)aminobenzoate)
Catalog # BC22
Supplier:  G-Biosciences
CAS Number:  
undefined
Sulpho-SIAB (Sulphosuccinimidyl (4-iodoacetyl)aminobenzoate)
Catalog # BC22
Supplier:  G-Biosciences
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Spacer Arm (Å):
    10.6
  • Reactive Toward:
    Primary Amine + Sulfhydryl
  • Membrane Permeable:
    No
  • Water Soluble:
    Yes
  • Cleavable/ Reversible:
    No

Specification Test Results

About this item

 :
 :504.19 g/mol
 :MFCD00054977
Formula:C₁₃H₁₀O₈N₂SINa