
SMPB (N-Succinimidyl 4-(4-Maleimidophenyl)butyrate)
Catalog # BC21
Supplier: G-Biosciences
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Spacer Arm (Å):11.6
- Reactive Toward:Primary Amine + Sulhydryl
- Membrane Permeable:Yes
- Water Soluble:No
- Cleavable/ Reversible:No
Specification Test Results
About this item
:
:356.33 g/mol
:GHS07
CAS Number:79886-55-8
Formula:C₁₈H₁₆N₂O₆
MDL Number:MFCD00054961