Order Entry
Northern Ireland
ContactUsLinkComponent
2,2'-Bipyridyl ≥99%, GPR RECTAPUR®
Toxic
2,2'-Bipyridyl ≥99%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Toxic
2,2'-Bipyridyl ≥99%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 99 %

Specification Test Results

Related Information

Frequently Bought Together