Sulpho-NHS-SS-Biotin, EZ-Link™, Pierce™
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:202057-28-1
:325143-98-4
Formula:C₁₉H₂₇O₉N₄S₄Na
MDL Number:MFCD00078533
:606.69 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Description:EZ-Link Sulfo-NHS-SS-Biotin
- Product Type:Sulfo-NHS-SS-Biotin
- Quantity:100 mg
- Product Line:EZ-Link™
- Cell Permeability:Cell-Impermeant
- Chemical Reactivity:Amine
- Reactive Moiety:Active Ester, Succinimidyl Ester, Sulfo-NHS Ester
- Solubility:DMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Water
- Label or Dye:Biotin
- Label Type:Biotin & Analogs
- Spacer:Mid-Length, Cleavable
Specification Test Results
Frequently Bought Together