Order Entry
Northern Ireland
ContactUsLinkComponent
Sulpho-SASD (Sulphosuccinimidyl 2-(4-azidosalicylamido)ethyl-1,3-dithiopropionate)
Sulpho-SASD (Sulphosuccinimidyl 2-(4-azidosalicylamido)ethyl-1,3-dithiopropionate)
Catalog # BC40
Supplier:  G-Biosciences
CAS Number:  
undefined
Sulpho-SASD (Sulphosuccinimidyl 2-(4-azidosalicylamido)ethyl-1,3-dithiopropionate)
Catalog # BC40
Supplier:  G-Biosciences
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :541.51 g/mol
CAS Number:144650-95-3
Formula:C₁₆H₁₆N₅NaO₉S₃
MDL Number:MFCD01861963