Order Entry
Northern Ireland
ContactUsLinkComponent
Sulpho-EMCS (N-(ε-maleimidocaproyloxy) sulphosuccinimide ester), Pierce™
Sulpho-EMCS (N-(ε-maleimidocaproyloxy) sulphosuccinimide ester), Pierce™
Catalog # PIER22307
CAS Number:  
undefined
Sulpho-EMCS (N-(ε-maleimidocaproyloxy) sulphosuccinimide ester), Pierce™
Catalog # PIER22307
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Seq:
    2370

Specifications

Specification Test Results

  • Product Line:
    Pierce™
  • Labeling Method:
    Chemical Labeling
  • Crosslinker Type:
    Heterobifunctional
  • Reactive Moiety:
    Maleimide, Sulfo-NHS Ester
  • Spacer:
    Short
  • Form:
    Powder
  • Quantity:
    50 mg
  • Type:
    Crosslinker
  • Solubility:
    Water

Specification Test Results

About this item

 :
 :408.32 g/mol
Storage Temperature:Refrigerator
Formula:C₁₄H₁₃O₉SN₂Na
MDL Number:MFCD09953186