SynaptoRed™ reagent ≥95% (by HPLC), Calbiochem®, Sigma-Aldrich®
Catalog # 574799-5
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Pack type:Plastic ampoule
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
:607.51 g/mol
:M-Clarity™ quality level = MQ100
Storage Temperature:Freezer
CAS Number:162112-35-8
Formula:C₃₀H₄₅Br₂N₃
MDL Number:MFCD00467819