PDPH ((3-(2-pyridyldithio)propionyl hydrazide)), Pierce™
Catalog # PIER22301
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:50 mg
- Seq:2370
Specifications
Specification Test Results
- Product Line:Pierce™
- Labeling Method:Chemical Labeling
- Crosslinker Type:Heterobifunctional
- Reactive Moiety:Maleimide, Pyridyldisulfide
- Spacer:Cleavable, Short
- Form:Powder
- Quantity:50 mg
- Type:Crosslinker
- Solubility:DMF, DMSO
Specification Test Results
About this item
:
:229.32 g/mol
Storage Temperature:Refrigerator
CAS Number:115616-51-8
Formula:C₈N₃S₂OH₁₁
MDL Number:MFCD00467204