ContactUsLinkComponent
CloneDetect
CloneDetect
Catalog # MLDVK8205
Supplier:  MOLECULAR DEVICES
CAS Number:  
CloneDetect
Catalog # MLDVK8205
Supplier:  MOLECULAR DEVICES
CAS Number:  
Specifications
  • Pk:
    1 Bottle
  • Description:
    Human IgG (Fc) specific, fluorescein
  • Seq:
    02
Specifications
About this item
Screen and select high value mammalian clones with label-free, in situ detection of IgG secretion with the ClonePix system, and other cell analysers with a green fluorescent channel.
  • Screen and select high-value mammalian clones with label-free, in situ detection of IgG secretion
  • A choice of detection agents specific for secreted mouse, rat or human IgGs
  • Sterile, virus and mycoplasma free
  • Optimised for ClonePix system, and compatible with cell analysers with a green fluorescent channel
A cell-culture validated, sterile and azide-free antibody reagent for detecting IgG in mammalian cell clone screening.