Cimetidine
ACRO467520050
:
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
- Appearance:White to almost white crystalline powder
- Infrared spectrum:Conforms
- Melting point:141°C to 143°C
- HPLC:≥ 97.5%
- Loss on drying:≤ 0.5%
:
:252.34 g/mol
:13.23
:141°C…143°C
:Ambient
:257-232-2
:51481-61-9
:C₁₀H₁₆N₆S
:MFCD01724314