ContactUsLinkComponent
Attachment for fire protection, b.safe
Attachment for fire protection, b.safe
  BOHLR580-05
 :  Bohlender
undefined
Attachment for fire protection, b.safe
  BOHLR580-05
 :  Bohlender
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    Attachment for fire protection, 134×135×38 mm, made of sheet metal, for connection of tube Ø max. 28 mm
  • Pk:
    1
Sheet metal case with gypsum insert for fire protection of cabinet or tube mounts in safety cabinets.
  • Includes 8 fastening screws
For fire protection of b.safe cabinet mounts GL 25 in safety cabinets.