Anti-RFP Polyclonal Antibody
ROCK200-101-379S
- Antigen symbol:RFP
- Clonality:Polyclonal
- Host:Goat
- Shipping temperature:Dry Ice
- Pk:25 µl
Goat anti-red fluorecent protein antibody
- Purified IgG Fraction
- No reaction was observed against human, mouse or rat serum proteins
- Western blot
Antibodies to RFP (Discosoma spp.) are intended for use in immunological assays including ELISA, western blotting, immunofluorescence, and fluorescence activated cell sorting (FACS).
Polyclonal anti-RFP is designed to detect RFP and its variants. Optimal titers for applications should be determined by the researcher. Expect a band ~27kDa in Western Blot in the appropriate cell lysate or extract.