Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-RANBP2 Rabbit polyclonal antibody
Anti-RANBP2 Rabbit polyclonal antibody
  NOVUNB100-93338
Anti-RANBP2 Rabbit polyclonal antibody
  NOVUNB100-93338

 

  • Antibody type:
    Primary
  • Antigen name:
    Ran-binding Protein 2
  • Antigen symbol:
    RanBP2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    5903
  • Antigen synonyms:
    RanBP2|P270|RAN binding protein 2|EC 5.2.1.8|ANE1|nucleoporin 358|Nuclear pore complex protein Nup358|E3 SUMO-protein ligase RanBP2|NUP358TRP2|TRP1|Ran-binding protein 2|transformation-related protein 2,358 kDa nucleoporin|Nucleoporin Nup358
  • Storage buffer:
    TBS and 0.1% BSA
  • Storage temperature:
    Store at 4 °C. Do not freeze.
  • Concentration:
    0,2 mg/ml
  • Immunogen:
    The immunogen recognized by this antibody maps to a region between residue 2600 and 2650 of human RAN binding protein 2 using the numbering given in entry NP_006258.3 (GeneID 5903).
  • Purification:
    Immunogen affinity purified
  • Pk:
    0,1 mL

 

 

The RanBP2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to RanBP2. This antibody reacts with human. The RanBP2 Antibody has been validated for the following applications: Western Blot, Immunoprecipitation.

Type: Primary
Antigen: RanBP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human