Anti-BRD4 Rabbit Polyclonal Antibody
Catalog # NOVUNBP1-86640
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen symbol:BRD4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Gene ID:23476
- Antigen synonyms:bromodomain containing 4|HUNK1bromodomain-containing protein 4|MCAP|CAPchromosome-associated protein|HUNKI|Protein HUNK1|bromodomain-containing 4
- Storage buffer:PBS (pH 7,2) and 40% Glycerol
- Storage temperature:Store at 4 °C short term. Aliquot and store at -20 °C long term. Avoid freeze-thaw cycles.
- Immunogen:This antibody was developed against Recombinant Protein corresponding to amino acids:PQPAKPQQVIQHHHSPRHHKSDPYSTGHLREAPSPLMIHSPQMSQFQSLTHQSPPQQNVQPKKQELRAASVVQPQPLVVVKEEKIHSPIIRSEPFSPSLRPEPPKHPESIKAPVHLPQRPEMKPVDVGRPVIRPPEQNAPPP
- Purification:Immunogen affinity purified
- Size:100 μl
- Pk:0,1 mL
Specifications
About this item
The BRD4 Antibody from Novus Biologicals is a rabbit polyclonal antibody to BRD4. This antibody reacts with human. The BRD4 Antibody has been validated for the following applications: Simple Western, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Simple Western
Type: Primary
Antigen: BRD4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human