Anti-ATF5 Goat Polyclonal Antibody
NOVUNBP1-51908
- Antibody type:Primary
- Antigen name:Activating Transcription Factor 5
- Antigen symbol:ATF5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:EVYKARSQRTRSC
- Gene ID:22809
- Antigen synonyms:Transcription factor ATFx|ATFXHMFN0395|cyclic AMP-dependent transcription factor ATF-5|cAMP-dependent transcription factor ATF-5|activating transcription factor 5FLJ34666
- Storage buffer:0,5 mg/ml Tris (pH 7,3) and 0,5% BSA
- Storage temperature:Store at –20 °C. Avoid freeze-thaw cycles.
- Immunogen:Peptide with sequence EVYKARSQRTRSC corresponding to C-Terminus according to NP_036200.2
- Purification:Immunogen affinity purified
- Size:100 μg
- Pk:0,1 mg
The ATF5 Antibody from Novus Biologicals is a goat polyclonal antibody to ATF5. This antibody reacts with human. The ATF5 Antibody has been validated for the following applications: Western Blot, Peptide ELISA.
Peptide
Type: Primary
Antigen: ATF5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: EVYKARSQRTRSC
Host: Goat
Isotype: IgG
Reactivity: Human