HIF-2 alpha overexpression lysate (adult normal)
Catalog # NOVUNBL1-10286
Supplier: Novus Biologicals
CAS Number:
Specifications
- Cell Format:Cryopreserved
- Pk:0,1 mg
- Tested applications:WB
Specifications
About this item
HIF-2 alpha overexpression lysate (adult normal).
Tested applications: Western blot
Tested reactivity: Human