ContactUsLinkComponent
Desktop mounting bracket for waste pipes, b.safe
Desktop mounting bracket for waste pipes, b.safe
Catalog # BOHLR1165-03
Supplier:  Bohlender
undefined
Desktop mounting bracket for waste pipes, b.safe
Catalog # BOHLR1165-03
Supplier:  Bohlender
Specifications
  • Description:
    Desktop mounting bracket for waste pipes, with silicone-free glue pad, fixing surface L×W: 260×44 mm, contact area L×W: 50×44 mm
  • Pk:
    1
Specifications
About this item
Contact area with silicone-free glue pad and 2 bores Ø 4,2 mm for non-slip fixing on the desktop.
  • Black, powder-coated angle plate made of steel
  • Compact collective connector to combine the outlets of two respectively up to four HPLC units
  • All connectors are placed sideways to join tubing and tubes with suitable fittings and hose connectors without bending
Waste Pipes with tube Ext.Ø 32 mm can be fixed and adjusted in height on the long side of the plate by using suitable b.safe Magnet Holders.