ContactUsLinkComponent
Magnet holder for waste pipes, b.safe
Magnet holder for waste pipes, b.safe
Catalog # BOHLR1162-32
Supplier:  Bohlender
undefined
Magnet holder for waste pipes, b.safe
Catalog # BOHLR1162-32
Supplier:  Bohlender
Specifications
  • Description:
    Magnet holder for waste pipes (Ø 32 mm), PE, L×W×H: 48×32×51 mm
  • Pk:
    1
Specifications
About this item
Pipe clamp made of PE with permanently fixed power magnets. b.safe Waste Pipes can easily be clipped into the pipe clamp for tube Ext.Ø 32 mm.
  • Compact collective connector to combine the outlets of two respectively up to four HPLC units
  • All connectors are placed sideways to join tubing and tubes with suitable fittings and hose connectors without bending
In combination with a suitable desktop- or wall mount, b.safe Waste Pipes can easily be fixed in height by means of the magnet.