Concentric reducer
Concentric reducer, 1,5×0,75''
Catalog # MSPPCOR61.5-0.75
Marketsource
Supplier: MARKET SOURCE PART PROCESS
Jump to:
Product Details & Documents
Manufactured from 316L pharmaceutical grade stainless steel, Pharma Hygiene Products’ concentric reducers provide the perfect solution for connecting existing Tri-Clamp connections to newer or changing ones.
- Durable
- Easy to sterilise and re-useable
- Crevice- and pit-free surfaces leave nowhere for bacteria to build up and store
- Finish is polished to an exceptional average finish of 0.8 micrometers Ra
Full material traceability and certification is available to meet international pharma quality compliance regulations.
Delivery information: Supplied complete with Tri-Clamp ferrule ends welded in place.
Specifications
- Description:Concentric reducer, 1,5×0,75''