Blank end caps
MSPPB21732012
- Description:Blank end caps, Ø 25 mm, for 0,5'' pipes
- Pk:1
Manufactured from 316L pharmaceutical grade stainless steel, Pharma Hygiene Products’ Tri-Clamp Blank Caps are designed to blank off Ferrule Ends, when used with Tri-Clamp gaskets and clamps.
- Durable
- Easy to sterilise and re-useable
- Crevice- and pit-free surfaces leave nowhere for bacteria to build up and store
- Can be cleaned with alcohol-based disinfectants, and in industrial dishwashers/autoclaves
Full material traceability is available to meet international pharma quality compliance regulations.
: Sold in packs of 10.