Polypropylene Bowl Covers
Keeps cleaned, non sterile production equipment surfaces protected from external sources of contamination; or use ‘in process’ to prevent cross contamination of products.
- Manufactured and packed in a controlled cleanroom environment
- Reduces labour costs
- Tear- and puncture-resistant non woven polypropylene
- Individually poly bagged
These polypropylene bowl covers are a great choice in non sterile pharmaceutical manufacturing operations. Can be used as a clean equipment cover or used 'in process' to prevent product cross contamination. Individually poly bagged to ensure cleanliness up until point of use.
: Certificates of Conformance sent with each shipment.
Frequently Bought Together