Viral inactivation solution, Ultrapure Bioreagent, J.T.Baker®
: Avantor
:9043-30-5
:0,9 – 1.1 g/ml
:Ambient
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:EPA safer choice chemical
- Identification (by FTIR):Passes Test
- Appearance:Passes Test
- Purity by HPLC:min 95%
- Assay:Passes Test
- Acid value:max 10
- Viscosity (25°C) :For information only
- Density:0.9 – 1.1 g/mL
- Limit of free ethylene oxide (Target <1 ppm):For information only
- Total microbial count (No growth detected):Passes Test
- Endotoxin:For information only
- Protein reactive impurities (100x Diluted):For information only
- Arsenic (As):For information only
- Cadmium (Cd):For information only
- Lead (Pb):For information only
- Mercury (Hg):For information only
- Cobalt (Co):For information only
- Nickel (Ni):For information only
- Vanadium (V):For information only
- Silver (Ag):For information only
- Gold (Au):For information only
- Iridium (Ir):For information only
- Osmium (Os):For information only
- Palladium (Pd):For information only
- Platinum (Pt):For information only
- Rhodium (Rh):For information only
- Ruthenium (Ru):For information only
- Selenium (Se):For information only
- Thallium (Tl):For information only
- Barium (Ba):For information only
- Chromium (Cr):For information only
- Copper (Cu):For information only
- Lithium (Li):For information only
- Molybdenum (Mo):For information only
- Antimony (Sb):For information only
- Tin (Sn):For information only
Frequently Bought Together