BM-1 orchid medium
Contains macro/micronutrients, vitamins and plantgrowth regulators required for orchid culturePlant tissue culture testedFull specification given in appendix.
- MFCD07370630
Contains macro/micronutrients, vitamins and plantgrowth regulators required for orchid culturePlant tissue culture testedFull specification given in appendix.
- MFCD07370630
Product Family Options
- DescriptionPk
- New ProductNew Product