- Pk:1
- Description:SureDirect blood PCR kit
- Seq:02
- Details:Specially formulated with a mutant of Taq DNA polymerase designed to overcome blood inhibitors for more robust performance. Designed for amplification of targets directly from whole blood and blood derivative samples.The 2X PCR master mix includes PCR enzyme, reaction buffer, magnesium and dNTPs
- No. of reactions:5×100 reactions
Enables direct DNA amplification from liquid blood, dried blood on paper, serum, plasma, and more.
- Streamlined workflow saves time
- No DNA Yield loss from DNA extraction
- Versatile: amplifies from many sample types, including liquid blood, frozen blood, dried blood on paper, serum, and plasma
- Outstanding robust performance: Multiplex up to three targets, good for high GC content (77%) and long amplicons (2,6 kb)
The SureDirect Blood PCR Kit enables direct DNA amplification from many sample types including: liquid blood, dried blood on paper, serum, and plasma. Inhibitor resistance and throughput, common PCR challenges associated with blood samples, are averted through the activity of a newly engineered Taq.
Your workflow is streamlined by eliminating the cumbersome DNA extraction step. SureDirect Blood PCR Kit’s outstanding robust performance will enable you to multiplex up to three targets and amplify high GC content (77%) and long amplicons (2,6 kb).