ContactUsLinkComponent
StrataPrep PCR purification kits, Agilent Technologies
StrataPrep PCR purification kits, Agilent Technologies
Catalog # AGLS400773
Supplier:  AGILENT
CAS Number:  
StrataPrep PCR purification kits, Agilent Technologies
Catalog # AGLS400773
Supplier:  AGILENT
CAS Number:  
Specifications
  • Pk:
    1
  • Description:
    StrataPrep PCR purification kit (250 reactions)
  • Seq:
    01
  • Sample size:
    250 reactions
  • Description:
    DNA-binding solution, PCR wash buffer, uspin cups and receptacle tubes.
Specifications
About this item
The StrataPrep PCR purification kits provide a rapid method to simultaneously purify PCR products from PCR primers, unincorporated nucleotides, buffer components, and enzymes.
  • Accommodates purification of PCR products from 100 bp to long PCR amplicons
  • Highly purified DNA, ready for use
  • No hazardous organic extraction or ethanol precipitation
  • Ideal purification for high throughput sequencing and microarrays
The StrataPrep PCR purification kits removes Contaminants such as Primers and Non-Specific Amplification Products From Reactions With Yields of Up To 10 µg. Purified DNA Exhibits Twice The Cloning Efficiency of Competitor Products.
Caution: For research use only. Not for use in diagnostic procedures.