Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Pk:1 KIT
- Application:RT-qPCR probe
- One step:Yes
- Description:SideStep II QRT-PCR master mix, 1-step
- No. of reactions:400
Complete kit for -step QRT-PCR from cells. Sufficient for 400 reactions.Use Brilliant QRT-PCR technology.
- Deliver efficient cell lysis
- RNA stabilization upto 6 months
- Sensitive quantitative gene expression analysis without RNA purification
- Purify the most relevant, interesting RNA samples
- Accurate data: RNA loss minimised
- Safe: No hazardous waste disposal and removal
These products are ideal for screening multiple samples first, saving more than 70% of the cost and 60% of the time it takes to column purify RNA.