Order Entry
Northern Ireland
ContactUsLinkComponent
NANOCOLOR® Photometric Tube Test Kit for Total Kjeldahl Nitrogen
NANOCOLOR® Photometric Tube Test Kit for Total Kjeldahl Nitrogen
Catalog # MANA985067
Supplier:  MACHEREY-NAGEL
CAS Number:  
undefined
NANOCOLOR® Photometric Tube Test Kit for Total Kjeldahl Nitrogen
Catalog # MANA985067
Supplier:  MACHEREY-NAGEL
CAS Number:  

Specifications

  • Chemical method:
    2,6 Dimethylphenol
  • Pk:
    1 KIT
  • Description:
    NANOCOLOR® total Kjeldahl Nitrogen TKN 16, test no. 0-67
  • Dimensions:
    343×166×137 mm (including packaging)
  • Model:
    MANA985067
  • Platform:
    NANOCOLOR® tube tests
  • Storage temperature:
    15…25 °C
  • Shelf life (months):
    12
  • Weight:
    1444 g including packaging
  • For unit:
    UV/VIS II, VIS II, 500 D, PF-12Plus, Advance, MT Easy VIS, MT Easy UV
  • Tests per kit:
    20
  • Range:
    1,00 - 16,0 mg/L (TKN)

Specifications

About this item

Photometric test kit for the determination of totoal Kjeldahl nitrogen in water samples.

  • Measurement directly in the test tube, saving time and reducing exposure to chemical reagents Bar code cuvette identification for easy test call up in NANOCOLOR® spectrophotometers Precisely pre-dosed reagents with constant high quality from batch to batch Coloured pictogram instructions and easy handling of the tubes 16 mm (OD) tubes for easy pipetting

The test kit is evaluated in 16 mm (OD) tubes. It is preprogrammend on NANOCOLOR® photometers and also compatible with METTLER TOLEDO Easy VIS and Easy UV. Programming data for other spectrophotometers are available.

Sea water analysis: No Hazardous material: Yes Remark: Measuring range on NANOCOLOR VIS II

Delivery: Supplied in 3 rugged boxes with 60 test tubes.