NANOCOLOR® Photometric Tube Test Kits for Chloride
Pre-dosed NANOCOLOR tube tests for photometric determination of chloride in water.
Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
- Barcode cuvette identification
- Precisely pre-dosed reagents in 16 mm (ext.Ø) tubes
The test kits are evaluated in 16 mm (ext.Ø) tubes. They are preprogrammend on NANOCOLOR® photometers. Programming data for other spectrophotometers are available.
Hazardous material: Yes
Remark: Measuring range on NANOCOLOR VIS II
: Rugged box with 20 test tubes.
- Model:MANA985019
- Platform:NANOCOLOR® tube tests
- Storage temperature:15…25 °C
- Shelf life (months):12
- Weight:455 g (including packaging)
Frequently Bought Together