Order Entry
Northern Ireland
ContactUsLinkComponent
5-(Difluoromethoxy)-2-mercaptobenzimidazole 99%
Irritant
5-(Difluoromethoxy)-2-mercaptobenzimidazole 99%
Catalog # APOSPC9818-25G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
5-(Difluoromethoxy)-2-mercaptobenzimidazole 99%
Catalog # APOSPC9818-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :216.21 g/mol
Melting Pt:239…243 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:97963-62-7
Formula:C₈H₆F₂N₂OS
MDL Number:MFCD00467504