Order Entry
Northern Ireland
ContactUsLinkComponent
1H-Undecafluoropentane 98%
Irritant
1H-Undecafluoropentane 98%
Catalog # APOSPC9755-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1H-Undecafluoropentane 98%
Catalog # APOSPC9755-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :270.05 g/mol
Boiling Pt:42 °C
Density:1,63 g/cm³ (25 °C)
 :GHS07
Storage Temperature:Ambient
CAS Number:375-61-1
Formula:C₅HF₁₁
MDL Number:MFCD08458172