3'-Fluoro-4'-nitroacetanilide 98%
Catalog # APOSPC909137-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:198.1512 g/mol
Storage Temperature:Ambient
CAS Number:345-30-2
Formula:C₈H₇FN₂O₃
MDL Number:MFCD01463345