Order Entry
Northern Ireland
ContactUsLinkComponent
Pentafluorophenyl trifluoroacetate 98%
Flammable
Irritant
Pentafluorophenyl trifluoroacetate 98%
Catalog # APOSPC8848-100G
Supplier:  Apollo Scientific
CAS Number:  
Flammable
Irritant
Pentafluorophenyl trifluoroacetate 98%
Catalog # APOSPC8848-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :280.07 g/mol
Boiling Pt:122…123 °C
Melting Pt:3 °C
Density:1,63 g/cm³ (25 °C)
Flash Pt:52 °C
UN:3272
ADR:3,III
 :GHS02
 :GHS07
Storage Temperature:Ambient
CAS Number:14533-84-7
Formula:C₈F₈O₂
MDL Number:MFCD00134438