
ABD-F [=4-(Aminosulphonyl)-7-fluoro-2,1,3-benzoxadiazole]
:91366-65-3
:
:C₆H₄FN₃O₃S
:MFCD00036687
:144…146 °C
:217.17 g/mol
:GHS07
:Freezer
Frequently Bought Together
Ethanol, GPR RECTAPUR®
: VWR Chemicals
Ethyl acetate ≥99%, GPR RECTAPUR®
: VWR Chemicals
Hexane (mixture of isomers), TECHNICAL
: VWR Chemicals
Water, TECHNICAL, demineralised
: VWR Chemicals
2-Propanol 70% ≥70% (v/v), GPR RECTAPUR®
: VWR Chemicals
Cyclohexane, EMPLURA®, Supelco®
: Merck
Ethylenediamine ≥99.0% (by GC), GPR RECTAPUR®
: VWR Chemicals
Toluene, anhydrous ≥99.8%, over molecular sieves
: Thermo Fisher Scientific
N,N-Dimethylformamide, extra dry over Molecular Sieve 99.8%, AcroSeal®
: Thermo Fisher Scientific