Trimethylsilyl trifluoromethanesulphonate 98%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: Trimethylsilyl trifluoromethanesulphonate
CAS Number:27607-77-8
:
Formula:C₄H₉F₃O₃SSi
MDL Number:MFCD00000406
Boiling Pt:37…40 °C/12mmHg
Density:1.228 g/cm³ (25 °C)
Molecular Weight:222.26 g/mol
Flash Pt:25 °C
Storage Temperature:Refrigerator
EINECS:248-565-4
ADR:3+8 III
Merck Index:12,09849
UN:2924
:GHS02
:GHS05
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together