Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(3,3,3-Trifluoropropyl)trimethoxysilane
(3,3,3-Trifluoropropyl)trimethoxysilane 97%
(3,3,3-Trifluoropropyl)trimethoxysilane 97%
APOSPC7826-25G
:
Apollo Scientific
:
(3,3,3-Trifluoropropyl)trimethoxysilane 97%
APOSPC7826-25G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
25 g
Seq:
3308
:
:
144 °C
:
1,137 g/cm³ (25 °C)
:
38 °C
:
218.25 g/mol
:
GHS02
:
Ambient
:
429-60-7
:
F₃CCH₂CH₂Si(OCH₃)₃
:
MFCD00039266