Order Entry
Northern Ireland
ContactUsLinkComponent
3-(Trifluoromethoxy)benzoyl chloride 97%
Corrosive
3-(Trifluoromethoxy)benzoyl chloride 97%
Catalog # APOSPC7438M-5G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
3-(Trifluoromethoxy)benzoyl chloride 97%
Catalog # APOSPC7438M-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :224.57 g/mol
Density:1,43 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:3265
ADR:8,II
 :GHS05
CAS Number:86270-03-3
Formula:C₈H₄ClF₃O₂
MDL Number:MFCD00061268