
3-(Trifluoromethoxy)benzonitrile 97%
Catalog # APOSPC7438F-10G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:187.12 g/mol
Melting Pt:37 °C
Density:1,31 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:3276
ADR:6.1,III
:GHS07
CAS Number:52771-22-9
Formula:C₈H₄F₃NO
MDL Number:MFCD00040959