
1H,1H,2H,2H-Perfluorooctanesulphonic acid 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:27619-97-2
:
Formula:C₈H₅F₁₃O₃S
MDL Number:MFCD00042455
:428.17 g/mol
Melting Pt:>300 °C (dec.)
Storage Temperature:Ambient
UN:3261
ADR:8,III
:GHS05
Frequently Bought Together