3-Fluoro-p-anisaldehyde
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:351-54-2
:
Formula:C₈H₇FO₂
MDL Number:MFCD00003349
:154.14 g/mol
Boiling Pt:129…132 °C/11mmHg
Melting Pt:34…35 °C
Flash Pt:>113 °C
Storage Temperature:Ambient
EINECS:206-514-3
:GHS07
Frequently Bought Together