


1-Butyl-3-methylimidazoliumtetrafluoroborate 98+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:174501-65-6
:
Formula:C₈H₁₅BF₄N₂
MDL Number:MFCD03095449
:226.02 g/mol
Melting Pt:–71 °C
Density:1,2077 g/cm³ (25 °C)
UN:3082
ADR:9+EHS,III
:GHS06
:GHS09
:GHS07
Storage Temperature:Ambient
Frequently Bought Together