(7-Azabenzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate (AOP) )
Catalog # APOSPC510112-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:443.27 g/mol
Storage Temperature:Ambient
CAS Number:156311-85-2
Formula:C₁₂H₂₂F₆N₆OP₂
MDL Number:MFCD09263289