
Lithium trifluoromethanesulphonate 97%
:33454-82-9
:
:CF₃LiO₃S
:MFCD00013227
:156.01 g/mol
:423 °C
:1,9 g/cm³ (25 °C)
:251-528-5
:GHS07
:Ambient
Frequently Bought Together
Cyclohexane, EMPLURA®, Supelco®
: Merck
Petroleum spirit, 40…60 °C, GPR RECTAPUR®
: VWR Chemicals
n-Pentane ≥95%, TECHNICAL
: VWR Chemicals
2-Propanol ≥99.0%, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Toluene ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Pyridine, extra dry over Molecular Sieve 99.5%, AcroSeal®
: Thermo Fisher Scientific
Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
: Honeywell Chemicals
N,N-Dimethylformamide, extra dry over Molecular Sieve 99.8%, AcroSeal®
: Thermo Fisher Scientific