1-Hexyl-3-methylimidazolium tetrafluoroborate
Catalog # APOSPC448020-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:254.08 g/mol
Storage Temperature:Ambient
CAS Number:244193-50-8
Formula:C₁₀H₁₉BF₄N₂
MDL Number:MFCD03095459