

(Difluoromethyl)trimethylsilane 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:65864-64-4
:
Formula:C₄H₁₀F₂Si
MDL Number:MFCD19381658
:124.21 g/mol
Boiling Pt:65…66 °C
Density:0,91 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:1993
ADR:3,II
:GHS02
:GHS07
Frequently Bought Together