
2,4-Dichloro-5-(trifluoromethyl)pyrimidine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:3932-97-6
:
Formula:C₅HCl₂F₃N₂
MDL Number:MFCD03426408
:216.98 g/mol
Boiling Pt:49 °C/32mmHg
Density:1,609 g/cm³ (25 °C)
Flash Pt:93 °C
Storage Temperature:Refrigerator
UN:2810
ADR:6.1,III
:GHS06
Frequently Bought Together